Learn More
WTW™ Cadmium Test Kits
Product Code. 11761073
Change view
Click to view available options
Range:
0.025 to 1 mg/L
Unit Size:
420g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
11761073 0.025 to 1 mg/L 420g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11761073 Supplier WTW™ Supplier No. 250314
 Restricted Item
In Stock
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Obtain precision measurements for waste water, seepage water, and process water with CombiCheck Cadmium (Cd) cell test kits.
  • For use with PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers
  • For waste water, seepage water, process water
  • CombiCheck Cadmium (Cd) cell test kit product type
  • Cadion derivative method
  • Photometric test results
TRUSTED_SUSTAINABILITY
Specifications
Analytes Cadion derivative
For Use With (Application) For waste water, seepage water, process water
For Use With (Equipment) PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers
Includes Certificates
No. of Tests 25
Range 0.025 to 1 mg/L
Sufficient For 25 Tests
Test Method Photometric
Type CombiCheck Cadmium (Cd) cell test kit
Product Identifier
  • Cadmium Cell Test Method: photometric 0.025 - 1.000 mg/l Cd Spectroquant
Supplemental information
  • MIXTURE LIST-Contain: Triton-X-100, disodium tetraborate; N,N-dimethylformamide