Learn More
WTW™ Cadmium Reagent Test Kits
Click to view available options
No. of Tests:
55
Unit Size:
Each
Description
- Convenient and cost-effective
- Assured quality by AQA/IQC
- For low concentrations and routine work with large sample throughput in the laboratory
- Wide measuring range using rectangular cuvettes for determination of trace concentrations
Specifications
Specifications
| Analytes | Rhodamine B |
| For Use With (Application) | For waste water, seepage water, process water |
| For Use With (Equipment) | For PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, spectroFlex photometers |
| Includes | Certificates |
| No. of Tests | 55 |
| Range | 0.002 - 0.500mg/L |
| Sufficient For | 55 Tests |
| Test Method | Photometric |
| Type | CombiCheck Gold (Au) reagent test kit |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu