Learn More
Cadmium Stearate, MP Biomedicals
Product Code. 11492266 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492266 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11492266 Supplier MP Biomedicals Supplier No. 0521273910
  • ONLINE EXCLUSIVE PRICE
    2160.00 NOK
    2055.93 NOK / 10g
    Save 104.07 NOK
    4% Off
 Restricted Item
In Stock
Acts as a lubricant and heat stabilizer in plastics.
Specifications
CAS 2223-93-0
Molecular Formula C36H70CdO4
Quantity 10 g
Synonym cadmium stearate, cadmium ii stearate, cadmium distearate, octadecanoic acid, cadmium salt, stabilisator scd, stabilizer scd, alaixol 11, cadmium octadecanoate, kadmiumstearat german, cadmium soaps stearate
InChI Key GWOWVOYJLHSRJJ-UHFFFAOYSA-L
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Cd+2]
IUPAC Name cadmium(2+);octadecanoate
Molecular Weight (g/mol) 679.366
PubChem CID 16681
Formula Weight 679.36
Grade Not Available
Physical Form Powder
Chemical Name or Material Cadmium Stearate