Learn More
Cadence Science™ Non-Coring Septum Penetration Needles
Needles allow for easier penetration and minimize coring and tracking
Brand:  Cadence Science™ 9862
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11726834
  • 17100.00 NOK / Pack of 12
Estimated Shipment: 02-10-2024
to see stock.
Description
Description
Needles allow for easier penetration and minimize coring and tracking
  • Micro-mate 316 stainless-steel hub
  • 316 stainless-steel tubing
Specifications
Specifications
Stainless Steel (Hub)
For easier penetration and minimize coring and tracking
Penetration Needle
25 mm
20 G
1 in.
12 Needles
Deflected End