Learn More
Cadence Science™ Non-Coring Septum Penetration Needles
Product Code. 11736834
Click to view available options
Needle Gauge:
18 G
20 G
Length (English) Needle:
1 in.
6 in.
8 in.
12 in.
Unit Size:
Pack of 12
Product Code. 11736834
Brand: Cadence Science™ 9863
Needles allow for easier penetration and minimize coring and tracking
  • Micro-mate 316 stainless-steel hub
  • 316 stainless-steel tubing
Specifications
Material Stainless Steel (Hub)
Needle Gauge 20 G
For Use With (Application) For easier penetration and minimize coring and tracking
Length (English) Needle 6 in.
Product Type Penetration Needle
Quantity 12 Needles
Needle Point Style Deflected End