Learn More
Cadence Science™ Laboratory Pipetting Needles
Product Code. 11764726
Click to view available options
Needle Gauge:
14 G
16 G
18 G
20 G
22 G
24 G
Length (English) Needle:
1 in.
1.5 in.
2 in.
4 in.
6 in.
Material:
Nickel-plated (Hub), Stainless Steel
Stainless Steel, Stainless Steel (Hub)
Connection Type:
Female Luer
Luer
Unit Size:
Pack of 12
Product Code. 11764726
Brand: Cadence Science™ 7931
316 Stainless Steel Hub with 316 Stainless Steel Tubing
  • Needles 14-825-16A through -16N and 14-818-106A through -106C all have standard nickel plated hubs and 304 stainless steel cannulae
  • Needles 01-290-29 through -35 have 316 stainless steel Micro-Mate™ Threaded Female Luer Hubs and 316 stainless steel cannulae
  • All have 90° blunt ends
Specifications
Material Nickel-plated (Hub), Stainless Steel
Needle Gauge 22 G
For Use With (Application) For both manual and automatic volumetric dispensers, diluters, pipetters and for laboratory procedures
Length (English) Needle 1.5 in.
Product Type Pipetting Needle
Quantity 12 Needles
Needle Point Style 90° Blunt
Connection Type Luer