Learn More
Cadence Science™ Laboratory Pipetting Needles
Product Code. 11705825
Change view
Click to view available options
Needle Gauge:
14 G
16 G
18 G
20 G
22 G
24 G
Length (English) Needle:
1 in.
1.5 in.
2 in.
4 in.
6 in.
Material:
Nickel-plated (Hub), Stainless Steel
Stainless Steel, Stainless Steel (Hub)
Connection Type:
Female Luer
Luer
Unit Size:
Pack of 12
Product Code. Needle Gauge Length (English) Needle Material Connection Type unitSize
11705825 14 G 4 in. Nickel-plated (Hub), Stainless Steel Luer Pack of 12
11774726 20 G 1.5 in. Nickel-plated (Hub), Stainless Steel Luer Pack of 12
11776824 16 G 4 in. Stainless Steel, Stainless Steel (Hub) Female Luer Pack of 12
11796824 14 G 4 in. Stainless Steel, Stainless Steel (Hub) Female Luer Pack of 12
11706834 14 G 6 in. Stainless Steel, Stainless Steel (Hub) Female Luer Pack of 12
11786824 16 G 6 in. Stainless Steel, Stainless Steel (Hub) Female Luer Pack of 12
11786814 20 G 4 in. Stainless Steel, Stainless Steel (Hub) Female Luer Pack of 12
11796814 18 G 2 in. Stainless Steel, Stainless Steel (Hub) Female Luer Pack of 12
11754726 24 G 1 in. Nickel-plated (Hub), Stainless Steel Luer Pack of 12
11764726 22 G 1.5 in. Nickel-plated (Hub), Stainless Steel Luer Pack of 12
11774936 18 G 2 in. Nickel-plated (Hub), Stainless Steel Luer Pack of 12
11715825 14 G 6 in. Nickel-plated (Hub), Stainless Steel Luer Pack of 12
11784936 18 G 4 in. Nickel-plated (Hub), Stainless Steel Luer Pack of 12
11764936 20 G 6 in. Nickel-plated (Hub), Stainless Steel Luer Pack of 12
11794936 18 G 6 in. Nickel-plated (Hub), Stainless Steel Luer Pack of 12
11785815 16 G 2 in. Nickel-plated (Hub), Stainless Steel Luer Pack of 12
11795815 16 G 4 in. Nickel-plated (Hub), Stainless Steel Luer Pack of 12
17 options
Product Code. 11705825
Brand: Cadence Science™ 7941
316 Stainless Steel Hub with 316 Stainless Steel Tubing
  • Needles 14-825-16A through -16N and 14-818-106A through -106C all have standard nickel plated hubs and 304 stainless steel cannulae
  • Needles 01-290-29 through -35 have 316 stainless steel Micro-Mate™ Threaded Female Luer Hubs and 316 stainless steel cannulae
  • All have 90° blunt ends
Specifications
Material Nickel-plated (Hub), Stainless Steel
Needle Gauge 14 G
For Use With (Application) For both manual and automatic volumetric dispensers, diluters, pipetters and for laboratory procedures
Length (English) Needle 4 in.
Product Type Pipetting Needle
Quantity 12 Needles
Needle Point Style 90° Blunt
Connection Type Luer