Learn More
CACYBP Antibody (7D11A8) [Janelia Fluor« 669], Novus Biologicals Biologicals™
Product Code. 30499902 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30499902 0.1 mL 0.1mL
1 options
Product Code. 30499902
Brand: Novus Biologicals NBP327912JF669
Mouse Monoclonal Antibody
CACYBP Monoclonal antibody specifically detects CACYBP in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Serum
TRUSTED_SUSTAINABILITY
Specifications
Antigen CACYBP
Applications ELISA, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7D11A8
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias CacyBP, calcyclin binding protein, GIG5, hCacyBP, MGC87971, RP1-102G20.6, S100A6-binding protein, S100A6BPgrowth-inhibiting gene 5 protein, Siah-interacting protein, Siah-interacting protein (SIP), SIPcalcyclin-binding protein
Host Species Mouse
Immunogen Truncated recombinant CACYBP expressed in E. Coli strain BL21 (DE3).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 27101
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1