missing translation for 'onlineSavingsMsg'
Learn More

Cacodylic acid, 98%

Product Code. p-7081894
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479972 2.5 g 2.5g
11489972 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11479972 Supplier Thermo Scientific Alfa Aesar Supplier No. A12075.88
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 75-60-5 | C2H7AsO2 | 138.00 g/mol

A useful arsenic acid for proteomics research. It a good substitute for phosphate in applications to avoid phosphates. Cacodylic Acid is useful for some DNA applications and is also popular in microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful arsenic acid for proteomics research. It a good substitute for phosphate in applications to avoid phosphates. Cacodylic Acid is useful for some DNA applications and is also popular in microscopy.

Solubility
Soluble in water (50 mg/ml), water (2000 at 25°C), alcohols, and acetic acid. Insoluble in ether.

Notes
Hygroscopic. Store away from moisture, air and oxidizing agents. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-60-5
Molecular Formula C2H7AsO2
Molecular Weight (g/mol) 138.00
MDL Number MFCD00002095
InChI Key OGGXGZAMXPVRFZ-UHFFFAOYSA-N
Synonym cacodylic acid, hydroxydimethylarsine oxide, chexmate, silvisar, phytar, arsan, dimethylarsenic acid, erase, arsinic acid, dimethyl, silvisar 510
PubChem CID 2513
ChEBI CHEBI:48765
IUPAC Name dimethylarsinic acid
SMILES C[As](C)(O)=O

Specifications

Melting Point 200°C
UN Number UN1572
Beilstein 1736965
Merck Index 14,1604
Solubility Information Soluble in water (50mg/ml),water (2000 at 25°C),alcohols,and acetic acid. Insoluble in ether.
Formula Weight 138
Percent Purity 98%
Quantity 2.5 g
Sensitivity Hygroscopic
Chemical Name or Material Cacodylic acid
compliance-icons compliance-icons
Product Identifier
  • Dimethylarsinic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: Cacodylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.