missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNA2D1 Rabbit anti-Human, Mouse, Porcine, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27453-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the alpha-2/delta subunit family, a protein in the voltage-dependent calcium channel complex. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Research on a highly similar protein in rabbit suggests the protein described in this record is cleaved into alpha-2 and delta subunits. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.Specifications
| CACNA2D1 | |
| Polyclonal | |
| Unconjugated | |
| CACNA2D1 | |
| CACNA2, CACNA2D1, CACNL2A, CCHL2A, MHS3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12293, 25399, 397377, 781 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O08532, P54289, P54290 | |
| CACNA2D1 | |
| CACNA2D1 Fusion Protein Ag26190 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction