missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNA1D Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22276-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Voltage-sensitive calcium channels (VSCC) mediate the entry of calcium ions into excitable cells and are also involved in a variety of calcium-dependent processes, including muscle contraction, hormone or neurotransmitter release, gene expression, cell motility, cell division and cell death. The isoform alpha-1D gives rise to L-type calcium currents. Long-lasting (L-type) calcium channels belong to the high-voltage activated (HVA) group. They are blocked by dihydropyridines (DHP), phenylalkylamines, benzothiazepines, and by omega-agatoxin-IIIA (omega-Aga-IIIA). They are however insensitive to omega-conotoxin-GVIA (omega-CTx-GVIA) and omega-agatoxin-IVA (omega-Aga-IVA).Specifications
CACNA1D | |
Polyclonal | |
Unconjugated | |
CACNA1D | |
CACH3, CACN4, CACNA1D, CACNL1A2, Cav1.3, CCHL1A2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
776 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q01668 | |
CACNA1D | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |