missing translation for 'onlineSavingsMsg'
Learn More

Cabazitaxel, Thermo Scientific™

Product Code. p-7238577 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 17585555

Brand: Thermo Scientific Acros 468891000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 183133-96-2
Molecular Formula C45H57NO14
Molecular Weight (g/mol) 835.94
MDL Number MFCD18827611
InChI Key BMQGVNUXMIRLCK-UHFFFAOYNA-N
IUPAC Name 4-(acetyloxy)-15-[(3-{[(tert-butoxy)carbonyl]amino}-2-hydroxy-3-phenylpropanoyl)oxy]-1-hydroxy-9,12-dimethoxy-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.0³,¹⁰.0⁴,⁷]heptadec-13-en-2-yl benzoate
SMILES COC1CC2OCC2(OC(C)=O)C2C(OC(=O)C3=CC=CC=C3)C3(O)CC(OC(=O)C(O)C(NC(=O)OC(C)(C)C)C4=CC=CC=C4)C(C)=C(C(OC)C(=O)C12C)C3(C)C

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Powder
Color White to Yellow to Beige
Melting Point 170°C to 180°C
Quantity 100 mg
Chemical Name or Material Cabazitaxel
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.