missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CA4 (Human) Recombinant Protein
Human CA4 full-length ORF ( AAH57792, 19 a.a. - 312 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000762-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: AESHWCYEVQAESSNYPCLVPVKWGGNCQKDRQSPINIVTTKAKVDKKLGRFFFSGYDKKQTWTVQNNGHSVMMLLENKASISGGGLPAPYQAKQLHLHWSDLPYKGSEHSLDGEHFAMEMHIVHEKEKGTSRNVKEAQDPEDEIAVLAFLVEAGTQVNEGFQPLVEALSNIPKPEMSTTMAESSLLDLLPKEEKLRHYFRYLGSLTTPTCDEKVVWTVFREPIQLHREQILAFSQKLYYDKEQTVSMKDNVRPLQQLGQRTVIKSGAPGRPLPWALPALLGPMLACLLAGFLR
Specifications
AAH57792 | |
carbonic anhydrase IV | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CA4 | |
GST |
762 | |
Wheat germ expression system | |
10 μg | |
CAIV, Car4, RP17 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |