missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CA125 VHH-8His-Cys-tag Recombinant Alpaca Monoclonal Antibody (SAA1355)

Product Code. 30131555
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30131555 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30131555 Supplier Invitrogen™ Supplier No. MA554644

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Alpaca Recombinant Monoclonal Antibody

Compatible with Direct ELISA.

CA 125 determinant is present on a high molecular weight, mucin like glycoprotein of high molecular weight. CA 125 has been found on frozen sections of amnion and derivatives of coelomic and mullerian epithelium, including pleura, pericardium and peritoneum. In adult tissues, epithelial cells of fallopian tube, endometrium and endocervix; pancreas, colon, gall bladder, stomach, kidney, apocrine sweat gland and mammary gland. It is also found in mesothelial cell lining of pleura, pericardium and peritoneum. It is found in ovarian tumors of serous, endometrioid or clear cell types and adenocarcinomas of mullerian type.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CA125 VHH-8His-Cys-tag
Applications ELISA, Surface Plasmon Resonance
Classification Recombinant Monoclonal
Clone SAA1355
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene MUC16
Gene Accession No. Q8WXI7
Gene Alias 1110008I14Rik; Ca125; CA-125; CA125 ovarian cancer antigen; ca-ovarian; FLJ14303; Gm1480; MUC16; MUC-16; mucin 16; mucin 16, cell surface associated; mucin-16; Ovarian cancer-related tumor marker CA125; ovarian carcinoma antigen CA125; RGD1308422
Gene Symbols MUC16
Host Species Alpaca
Immunogen Recombinant Human MUC16/CA125.
Purification Method Metal-chelate chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94025
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype VHH
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.