Learn More
C9-12 Isoalkanes, TRC
Specifications
Specifications
| Chemical Name or Material | Isopar G |
| CAS | 90622-57-4 |
| Quantity | 500 mL |
| Synonym | C9-12-iso-alkanes, Isopar H, Isopar K, UltraClear, Isopar G |