missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C8orf41 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20779-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The DNA replication (origin) licensing system is necessary for duplicating the genome in each cell cycle. Geminin is a protein that prevents DNA replication by blocking the MCM complex incorporation into prereplication complex (pre-RC). It is absent during G1 phase and accumulates during S, G2, and M phases of mitosis. It is then degraded during the metaphase-anaphase trasition allowing replication to occur during subsequent cycles. Geminin has also been implicated in maintaining integrity of the male germline genome.Specifications
C8orf41 | |
Polyclonal | |
Unconjugated | |
TTI2 | |
C8orf41, TTI2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
80185 | |
-20°C | |
Liquid |
Western Blot | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6NXR4 | |
TTI2 | |
C8orf41 Fusion Protein Ag14776 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |