missing translation for 'onlineSavingsMsg'
Learn More

C8orf41 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20779-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16890345

  • 1611.84 NOK / 20µL
Estimated Shipment: 08-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The DNA replication (origin) licensing system is necessary for duplicating the genome in each cell cycle. Geminin is a protein that prevents DNA replication by blocking the MCM complex incorporation into prereplication complex (pre-RC). It is absent during G1 phase and accumulates during S, G2, and M phases of mitosis. It is then degraded during the metaphase-anaphase trasition allowing replication to occur during subsequent cycles. Geminin has also been implicated in maintaining integrity of the male germline genome.
Specifications

Specifications

C8orf41
Polyclonal
Unconjugated
TTI2
C8orf41, TTI2
Rabbit
Antigen Affinity Chromatography
RUO
80185
-20°C
Liquid
Western Blot
0.46 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6NXR4
TTI2
C8orf41 Fusion Protein Ag14776
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers