missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C7orf43 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27790-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The predicted MW of C7orf43 is 62 kDa. 27790-1-AP recognizes a 65-70 kDa band possibly due to phosphorylation.
The function of this protein remains unknown.Specifications
C7orf43 | |
Polyclonal | |
Unconjugated | |
C7orf43 | |
C7orf43 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
288545, 55262 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8WVR3 | |
C7orf43, RGD1305455 | |
C7orf43 Fusion Protein Ag27149 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |