Description
ARMT1 gene ontology annotations related to this gene include protein carboxyl O-methyltransferase activity.
Specifications
Specifications
| Antigen | C6orf211 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 0.22 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | ARMT1 |
| Gene Accession No. | Q9H993 |
| Gene Alias | C6orf211, UPF0364 protein C6orf211 |
| Gene Symbols | ARMT1 |
| Show More |