Learn More
C6orf150 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26416-1-AP-150UL
This item is not returnable.
View return policy
Description
The exact function of C6orf150 remains unknown.Specifications
| C6orf150 | |
| Polyclonal | |
| Unconjugated | |
| CGAS | |
| C6orf150, cGAS, Cyclic GMP-AMP synthase, h-cGAS, MB21D1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 115004 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8N884 | |
| CGAS | |
| C6orf150 Fusion Protein Ag23455 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |