missing translation for 'onlineSavingsMsg'
Learn More

C6orf15 Rabbit anti-Human, Non-human primate, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 24953-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16873475

  • 4593.76 NOK / 150µL
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

C6orf15, also named as Protein STG, is a 325 amino acid protein, which as a secreted protein is expressed in skin and tonsils. The calculated molecular weight of C6orf15 is 34 kDa, but we detected a 50-55 kDa protein by western blot. C6orf15 may contains some modified residues.
Specifications

Specifications

C6orf15
Polyclonal
Unconjugated
C6orf15
C6orf15, Protein STG, STG
Rabbit
Antigen Affinity Chromatography
RUO
29113
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.28 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6UXA7
C6orf15
C6orf15 Fusion Protein Ag20276
150 μL
Primary
Human, Non-human Primate
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers