Learn More
C6orf108 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25596-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
RCL was identified on the basis of its stimulation by c-Myc protein. The latter is a transcription factor that participates in the regulation of cell proliferation, differentiation, and apoptosis. The exact function of this gene is not known but studies in rat suggest a role in cellular proliferation and c-Myc-mediated transformation.Specifications
C6orf108 | |
Polyclonal | |
Unconjugated | |
DNPH1 | |
c Myc responsive protein Rcl, C6orf108, RCL, RP3 330M21.3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10591 | |
-20°C | |
Liquid |
Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43598 | |
DNPH1 | |
C6orf108 Fusion Protein Ag22336 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |