Learn More
C6orf108 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25596-1-AP-20UL
 View more versions of this product
Product Code. 16804165
  • 1710.33 NOK / 20µL
Description
RCL was identified on the basis of its stimulation by c-Myc protein. The latter is a transcription factor that participates in the regulation of cell proliferation, differentiation, and apoptosis. The exact function of this gene is not known but studies in rat suggest a role in cellular proliferation and c-Myc-mediated transformation.
Specifications
Specifications
C6orf108
Polyclonal
Unconjugated
DNPH1
c Myc responsive protein Rcl, C6orf108, RCL, RP3 330M21.3
Rabbit
Antigen Affinity Chromatography
RUO
10591
-20°C
Liquid
Western Blot
0.34 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O43598
DNPH1
C6orf108 Fusion Protein Ag22336