missing translation for 'onlineSavingsMsg'
Learn More

C5 Mouse anti-Human, Clone: 2G8C9, Proteintech

Product Code. 16848695 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16848695 20 μL 20µL
16838695 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16848695

Brand: Proteintech 666341IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

C5 is part of the membrane attack complex (MAC) of the complement system. C5 is split into C5a og C5b by the C5 convertases from the classical and the alternative complement pathway and the C5b fragment initiates the assembly of the terminal MAC (C5b-C9). The molecular mass of C5 is 190 kDa.

Specifications

Antigen C5
Applications Western Blot
Classification Monoclonal
Clone 2G8C9
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene C5
Gene Accession No. P01031
Gene Alias C5, Complement C5, complement component 5, CPAMD4
Gene Symbols C5
Host Species Mouse
Immunogen C5 Fusion Protein Ag17882
Purification Method Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 727
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.