Learn More
C3orf64 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27595-1-AP-150UL
 View more versions of this product
Product Code. 16895715
  • 4836.10 NOK / 150µL
Specifications
C3orf64
Polyclonal
Unconjugated
EOGT
AER61, C3orf64, EGF domain-specific O-linked GlcNAc transferase, EOGT, EOGT1
Rabbit
Antigen Affinity Chromatography
RUO
101351, 285203
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5NDL2, Q8BYW9
EOGT
C3orf64 Fusion Protein Ag25932
150 μL
Primary
Human, Mouse