Learn More
C2orf37 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26033-1-AP-20UL
 View more versions of this product
Product Code. 16804565
  • 1710.33 NOK / 20µL
Specifications
C2orf37
Polyclonal
Unconjugated
DCAF17
C2orf37, DCAF17
Rabbit
Antigen Affinity Chromatography
RUO
75763, 80067
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q3TUL7, Q5H9S7
Dcaf17
C2orf37 Fusion Protein Ag21995
20 μL
Primary
Mouse, Human