missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C22orf33 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26577-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| C22orf33 | |
| Polyclonal | |
| Unconjugated | |
| TEX33 | |
| C22orf33, cE81G9.2, EAN57, Protein EAN57 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 339669 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O43247 | |
| TEX33 | |
| C22orf33 Fusion Protein Ag23900 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction