missing translation for 'onlineSavingsMsg'
Learn More

C22orf33 Rabbit anti-Human, Polyclonal, Proteintech

Product Code. 16834995 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16834995 150 μL 150µL
16844995 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16834995

Brand: Proteintech 265771AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Specifications

Antigen C22orf33
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TEX33
Gene Accession No. O43247
Gene Alias C22orf33, cE81G9.2, EAN57, Protein EAN57
Gene Symbols TEX33
Host Species Rabbit
Immunogen C22orf33 Fusion Protein Ag23900
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339669
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.