missing translation for 'onlineSavingsMsg'
Learn More

C21orf91 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

3571.05 NOK - 5364.45 NOK

Specifications

Antigen C21orf91
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18208884
View Documents
Novus Biologicals
NBP2-55022
100 μL
ONLINE EXCLUSIVE PRICE
5675.00 NOK
5364.45 NOK / 100µL
Save 310.55 NOK
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18615417
View Documents
Novus Biologicals
NBP2-55022-25ul
25 μL
ONLINE EXCLUSIVE PRICE
3780.00 NOK
3571.05 NOK / 25µL
Save 208.95 NOK
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

C21orf91 Polyclonal specifically detects C21orf91 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

C21orf91
Polyclonal
Rabbit
Human
C21orf14, C21orf38, C21orf7, chromosome 21 open reading frame 38, chromosome 21 open reading frame 91, CSSG1, DKFZp781D1223, early undifferentiated retina and lens, EURL, protein EURL homolog, YG81, YG81C21orf14
C21ORF91
IgG
Affinity Purified
Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
54149
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:EEEQFVNIDLNDDNICSVCKLGTDKETLSFCHICFELNIEGVPKSDLLHTKSLRGHKDCFEKYHLIANQGCPRSKLS
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.