missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1QBP Mouse anti-Human, Mouse, Clone: 1F9B1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68084-1-IG-150UL
This item is not returnable.
View return policy
Description
C1QBP Monoclonal antibody apecifically detetects C1QBP antigen in Human, Mouse Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.Specifications
| C1QBP | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| C1QBP | |
| C1QBP fusion protein Ag19773 | |
| 150 μL | |
| 708 | |
| Store at -20°C. Stable for one year after shipment. | |
| IgG2b |
| Western Blot, Immunohistochemistry, ELISA | |
| 1F9B1 | |
| Unconjugated | |
| BC013731 | |
| Mouse | |
| Protein A purification | |
| Primary | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction