missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1qA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11602-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is raised against C1qA which is the A-chain polypeptide of human complement subcomponent C1q.
C1q is a sub-component of complement C1. C1q associates with complement components C1r and C1s, which are required for the progression of the proteolytic complement cascade. Deficiency of C1q has been associated with lupus erythematosus and glomerulonephritis. C1q is composed of 18 polypeptide chains: six A-chains, six B-chains, and six C-chains. Each chain contains a collagen-like region located near the N terminus and a C-terminal globular region. The A-, B-, and C-chains are arranged in the order A-C-B on chromosome 1.Specifications
| C1qA | |
| Polyclonal | |
| Unconjugated | |
| C1QA | |
| C1qA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12259, 298566, 712 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P02745, P31720, P98086 | |
| C1QA | |
| C1qA Fusion Protein Ag2141 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction