Learn More
C1QA Mouse anti-Human, Unconjugated, Clone: 1B10E3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67063-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The C1QA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human C1QA. This antibody recognizes human antigen. The C1QA antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
C1QA | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
1B10E3 | |
Unconjugated | |
20 μL | |
712 | |
IgG1 |