missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C1q Monoclonal Antibody (JL-1), FITC
GREENER_CHOICE

Product Code. 10613504
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Product Code. Quantity unitSize
10613504 100 μg 100µg
1 options
This item is not returnable. View return policy

Product Code. 10613504

Brand: Invitrogen™ MA140313

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-40313 detects C1q from mouse, rat, and human samples. MA1-40313 has been successfully used in immunofluorescence, immunohistochemistry (frozen), ELISA, functional assays and Western blot applications. The MA1-40313 immunogen is Purified Mouse C1q. Positive control: Spleen and kidney tissue of wild-type mice; Negative control: Spleen and kidney tissue of C1q -/- mice.

C1q is a sub-component of complement C1. C1q associates with complement components C1r and C1s, which are required for the progression of the proteolytic complement cascade. Deficiency of C1q has been associated with lupus erythematosus and glomerulonephritis. C1q is composed of 18 polypeptide chains: six A-chains, six B-chains, and six C-chains. Each chain contains a collagen-like region located near the N terminus and a C-terminal globular region. The A-, B-, and C-chains are arranged in the order A-C-B on chromosome 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1q
Applications ELISA, Functional Assay, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone JL-1
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 1% BSA and 0.02% sodium azide
Gene C1QA
Gene Accession No. P02745, P02746, P02747, P14106, P31720, P31721, P31722, P98086, Q02105
Gene Alias AI255395; AI385742; C1q; C1qa; C1qb; C1qc; C1Q-C; C1QG; Ciqc; complement C1q A chain; complement C1q B chain; complement C1q C chain; complement C1q chain A; complement C1q chain B; complement C1q subcomponent subunit A; complement C1q subcomponent subunit A-like protein; Complement C1q subcomponent subunit B; Complement C1q subcomponent subunit C; complement component 1, q subcomponent, A chain; complement component 1, q subcomponent, alpha polypeptide; complement component 1, q subcomponent, B chain; complement component 1, q subcomponent, beta polypeptide; complement component 1, q subcomponent, C chain; complement component 1, q subcomponent, c polypeptide; complement component 1, q subcomponent, gamma polypeptide; complement component C1q, A chain; complement component C1q, B chain; complement subcomponent C1q chain B
Gene Symbols C1QA, C1QB, C1qc
Host Species Mouse
Immunogen Purified Mouse C1q.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12259, 12260, 12262, 29687, 298566, 362634, 712, 713, 714
Target Species Human, Mouse, Rat
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.