missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26074-1-AP-150UL
Additional Details : Weight : 0.00010kg
Specifications
C1orf9 | |
Polyclonal | |
Unconjugated | |
SUCO | |
C1orf9, CH1, OPT, Protein osteopotentia homolog, SUCO | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
226551, 51430 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8C341, Q9UBS9 | |
SUCO | |
C1orf9 Fusion Protein Ag23224 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |