Learn More
C1orf226 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26061-1-AP-150UL
Additional Details : Weight : 0.00010kg
Specifications
C1orf226 | |
Polyclonal | |
Unconjugated | |
C1orf226 | |
C1orf226 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
400793 | |
-20°C | |
Liquid |
Immunoprecipitation | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A1L170 | |
C1orf226 | |
C1orf226 Fusion Protein Ag21811 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |