missing translation for 'onlineSavingsMsg'
Learn More

C1orf163 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16843945 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16843945 20 μL 20µL
16833945 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16843945

Brand: Proteintech 253611AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Required for assembly of mitochondrial respiratory chain complex I and complex IV.

Specifications

Antigen C1orf163
Applications Immunofluorescence, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene COA7
Gene Accession No. Q921H9, Q96BR5
Gene Alias C1orf163, COA7, RESA1, Sel1 repeat-containing protein 1, SELRC1
Gene Symbols Coa7
Host Species Rabbit
Immunogen C1orf163 Fusion Protein Ag19734
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65260, 69893
Target Species Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.