Learn More
C1orf116 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Product Code. 16893774 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16893774 150 μL 150µL
16803784 20 μL 20µL
2 options
Product Code. 16893774
Brand: Proteintech 148881AP150UL
Rabbit Polyclonal Antibody
Two isoforms of SARG are produced by alternative splicing encoding 64 kDa and 38 kDa proteins, and both isoforms can be recognized by this antibody (Product No. 14888-1-AP).
Specifically androgen-regulated gene protein (SARG), encoded by C1orf116, is a 601-aa protein considered putative androgen-specific receptor. SARG mRNA is highly expressed in prostate and can be up-regulated by androgens, but not by glucocorticoids. Little is known about the function of SARG to date.
Specifications
Antigen C1orf116
Applications Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SARG
Gene Accession No. Q9BW04
Gene Alias C1orf116, SARG
Gene Symbols AA986860, SARG
Host Species Rabbit
Immunogen C1orf116 Fusion Protein Ag6675
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 212439, 79098
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG