missing translation for 'onlineSavingsMsg'
Learn More

C188-9, MedChemExpress

Product Code. 16433166
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16433166 25 mg 25mg
16475456 100 mg 100mg
16457327 5 mg 5mg
16404618 10 mg 10mg
16400917 50 mg 50mg
5 options
This item is not returnable. View return policy

Product Code. 16433166

Brand: Medchem Express HY112288

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress C188-9 (TTI-101) is a STAT3 inhibitor, with a Kd of 4.7 nM. C188-9 inhibits G-CSF-induced STAT3 activation and STAT3-dependent gene expression. C188-9 induces apoptosis in AML cell lines and primary samples and inhibits colony formation by primary AML blasts.

C188-9 (TTI-101) is a STAT3 inhibitor, with a Kd of 4.7 nM. C188-9 inhibits G-CSF-induced STAT3 activation and STAT3-dependent gene expression. C188-9 induces apoptosis in AML cell lines and primary samples and inhibits colony formation by primary AML blasts.

Specifications

Chemical Name or Material C188-9
CAS 432001-19-9
Color Pink
Purity Grade Notes Research
Molecular Formula C27H21NO5S
Quantity 25 mg
Synonym TTI-101
Solubility Information DMSO : 25 mg/mL (53.02 mM; ultrasonic and warming and heat to 60°C)
SMILES O=S(C1=CC=C(OC)C=C1)(NC2=C3C=CC=CC3=C(O)C(C4=C5C=CC=CC5=CC=C4O)=C2)=O
Molecular Weight (g/mol) 471.52
Formula Weight 471.52
Percent Purity 98.11%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.