Learn More
Macherey-Nagel™ C18 Ec F Adsorbent
Product Code. 10134661 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10134661 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10134661 Supplier Macherey-Nagel™ Supplier No. 730613
  • ONLINE EXCLUSIVE PRICE
    3565.00 NOK
    3281.38 NOK / 100g
    Save 283.62 NOK
    7% Off
 Restricted Item
Estimated Shipment: 28-12-2026