missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C13orf27/TEX30 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25408-1-AP-20UL
This item is not returnable.
View return policy
Description
C13orf27 is a protein coding gene.Specifications
| C13orf27/TEX30 | |
| Polyclonal | |
| Unconjugated | |
| TEX30 | |
| C13orf27, TEX30 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 75623, 93081 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q3TUU5, Q5JUR7 | |
| Tex30 | |
| C13orf27/TEX30 Fusion Protein Ag22096 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction