Learn More
C11orf80 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25957-1-AP-20UL
 View more versions of this product
Product Code. 16804495
  • 1710.33 NOK / 20µL
Description
C11orf80 locates at the chromosome 11. It has two isoforms with MW 70-75 kDa.
Specifications
Specifications
C11orf80
Polyclonal
Unconjugated
TOP6BL
C11orf80
Rabbit
Antigen Affinity Chromatography
RUO
79703
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8N6T0
TOP6BL
C11orf80 Fusion Protein Ag22895