Learn More
C11orf70 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Product Code. 16800355 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16800355 150 μL 150µL
16810355 20 μL 20µL
2 options
Product Code. 16800355
Brand: Proteintech 207801AP150UL
Rabbit Polyclonal Antibody
Specifications
Antigen C11orf70
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene C11orf70
Gene Accession No. Q9BRQ4
Gene Alias CK070, C11orf70
Gene Symbols C11orf70
Host Species Rabbit
Immunogen C11orf70 Fusion Protein Ag14793
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85016
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG