missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Involved in redox regulation of the cell. Acts as an antioxidant. Inhibits TNFSF11-induced NFKB1 and JUN activation and osteoclast differentiation. May affect bone resorption and help to maintain bone mass.
Specifications
Specifications
| Antigen | C10orf58 |
| Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.26 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | FAM213A |
| Gene Accession No. | Q6AXX6, Q9BRX8 |
| Gene Alias | C10orf58, F213A, FAM213A, PSEC0139, UNQ611/PRO1198, UPF0765 protein C10orf58 |
| Gene Symbols | Fam213a |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?