Learn More
Motic™ C-Mount Camera Adapter for SMZ-160
Product Code. 17827295
Change view
Click to view available options
Magnification Power:
0.35X
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Magnification Power Unit Size
17827295 0.35X Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17827295 Supplier Motic™ Supplier No. 1101010100861
  • ONLINE EXCLUSIVE PRICE
    1380.00 NOK
    1202.31 NOK / Pack of 1
    Save 177.69 NOK
    12% Off
Estimated Shipment: 14-12-2026
The C-Mount Camera Adapter is compatible with the SMZ-160 microscope.
  • C-mount camera adapter for SMZ-160.
  • Available in 0.35x, 0.5x, or 1.0x (no lens) magnification.
  • Adapt 1/3 in. or 1/2 in. chip sensors.
Specifications
Type Adapter
Autoclavable Not Autoclavable
Camera Type C-Mount
For Use With (Application) To screw a digital camera on a trinocular SMZ-160
For Use With (Equipment) To be used with a digital camera for microscopy
Magnification Power 0.35X
Warranty 5 Years