Description
- UL, CSA and CE standards certified
- Used for adherent cell cultures
- For laboratory, pilot plant and production laboratory cell culture installations
Specifications
Specifications
| Speed Range | 0.25 to 8.1 rpm |
| Electrical Requirements | 100/240 V, 50/60 Hz, 35 W |
| Voltage | 100/240 V |
| Temperature Range (Metric) | 10°C to 40°C |
| Wattage | 35 W |
| Certifications/Compliance | CE, CSA, UL |
| Frequency | 50/60 Hz |
| Model | C Model |
| For Use With (Application) | For Adherent Cell Culture |
| Humidity Range | 80% up to 31°C |
| Show More |