Learn More
Wheaton™ C Model R2P 2.0 Roller Culture Apparatus
Product Code. 13403886 Shop All Wheaton Science Products Products
Click to view available options
No. of Positions:
88
Unit Size:
Each
Product Code. 13403886
Brand: Wheaton™ WRBPF8110C
Only null left
Specifications
Speed Range 0.25 to 8.1 rpm
Electrical Requirements 100/240 V, 50/60 Hz, 35 W
Voltage 100/240 V
Temperature Range (Metric) 10°C to 40°C
Wattage 35 W
Certifications/Compliance CE, CSA, UL
Frequency 50/60 Hz
Depth (Metric) 63 cm
Height (Metric) 187 cm
Width (Metric) 120 cm
Weight (Metric) 167 kg
Model C Model
For Use With (Application) For Adherent Cell Culture
Humidity Range 80% up to 31°C
No. of Positions 88
Packaging Quantity 1 Ea.
Spacing 15.2 in.