Learn More
Wheaton™ C Model Bottom Drive Modular Spacing Roller Culture Apparatus
Product Code. 15204568 Shop All Wheaton Science Products Products
Change view
Click to view available options
Includes:
1 Deck
5 Decks
9 Decks
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
15204568 5 Decks Each
13433896 9 Decks Each
15214467 1 Deck Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15204568 Supplier Wheaton™ Supplier No. WSBMR5050C
 Restricted Item
Estimated Shipment: 29-12-2026
Specifications
Speed Range 0.25 to 8.1 rpm
Electrical Requirements 100/240 V, 50/60 Hz, 35 W
Voltage 100/240 V
Temperature Range (Metric) 10°C to 40°C
Wattage 35 W
Certifications/Compliance CE, CSA, UL
Frequency 50/60 Hz
Model C Model
For Use With (Application) For laboratory, pilot plant and production cell culture installations
Humidity Range 80% up to 31°C
Includes 5 Decks
Packaging Quantity 1 Ea.
Spacing 17.7 cm