Learn More
c-Kit-IN-1, MedChemExpress
Product Code. 16466388
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16466388
Brand: Medchem Express HY15240
MedChemExpress c-Kit-IN-1 is a potent inhibitor of c-Kit and c-Met with IC50s of <200 nM.
c-Kit-IN-1 is a potent inhibitor of c-Kit and c-Met with IC50s of <200 nM.
Specifications
Chemical Name or Material c-Kit-IN-1
CAS 1225278-16-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H21F2N5O3
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (204.30 mM)
SMILES O=C(C1(CC1)C(NC2=CC=CC=C2)=O)NC3=CC(F)=C(OC4=CC=NC(C5=CN(C)N=C5)=C4)C=C3F
Molecular Weight (g/mol) 489.47
Formula Weight 489.47
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.