Learn More
Masterflex™ Transfer Tubing, C-Flex™, Opaque White
Product Code. p-8003607
Change view
Click to view available options
Diameter (Metric) Inner:
0.51 mm
1.6 mm
3 mm
4.8 mm
6.4 mm
7.9 mm
9.6 mm
Diameter (Metric) Outer:
2.1 mm
2.4 mm
3.2 mm
6.4 mm
9.6 mm
11.1 mm
12.7 mm
15.9 mm
Maximum Pressure:
1 bar
10 psi
12 psi
14 psi
16 psi
17 psi
2 bar
21 psi
22 psi
31 psi
9 psi
Unit Size:
7.5m
Each
Product Code. Diameter (Metric) Inner Diameter (Metric) Outer Maximum Pressure unitSize
11717529 3 mm 6.4 mm 9 psi 7.5m
11783149 4.8 mm 11.1 mm 22 psi Each
11797509 4.8 mm 2.4 mm 31 psi 7.5m
11757529 7.9 mm 11.1 mm 10 psi 7.5m
11767529 7.9 mm 12.7 mm 16 psi 7.5m
11777529 9.6 mm 12.7 mm 10 psi 7.5m
11787529 9.6 mm 15.9 mm 14 psi 7.5m
11787509 0.51 mm 2.1 mm 2 bar 7.5m
11727529 6.4 mm 9.6 mm 12 psi 7.5m
11707519 1.6 mm 3.2 mm 1 bar 7.5m
11797519 3 mm 6.4 mm 21 psi 7.5m
11747529 6.4 mm 12.7 mm 17 psi 7.5m
12 options
Product Code. 11717529
Brand: Masterflex™ 0642466
Only null left
Add to basket
Add to basket
Meets the demands of various industries
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
Specifications
Certifications/Compliance Resin meets USP Class VI requirements. Tubing is manufactured according to GMP
Autoclavable Autoclavable
Diameter (Metric) Inner 3 mm
Material C-Flex
Diameter (Metric) Outer 6.4 mm
Overall Length (Metric) 7.62 m
For Use With (Application) General-purpose lab uses requiring high performance and long life
Min. Operating Temperature (Metric) -45°C
Packaging Quantity 1 Pack
Sterility Sterile
Color White
Maximum Pressure 9 psi
Max. Operating Temperature (Metric) 135°C