Learn More
Masterflex™ C-Flex™ L/S™ Spooled Precision Pump Tubing
Click to view available options
Tubing Size:
13
14
15
16
17
18
24
25
35
36
Flow Rate:
0.21 to 130mL/min.
1000 mL/min
1100 mL/min
1700 mL/min
1800 mL/min
2300 mL/min.
2600 mL/min
3400 mL/min
36 mL/min
480 mL/min
Conditionnement:
Each
Description
- Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique application
- Always in stock—most formulations and sizes available to ship same day
- Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
- Very low protein binding
- Heat sealable, weldable
- Economical tubing
- Applications: General-purpose lab uses requiring high performance and long life
- Characteristics: Exceptional tensile and tear strength, good biocompatibility, low gas permeability, smooth surface, opaque white
- Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils
Spécification
Spécification
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 0.8 mm |
| Material | C-Flex |
| For Use With (Application) | For General-Purpose Lab Uses Requiring High Performance and Long Life |
| Tubing Size | 13 |
| Flow Rate | 36 mL/min |
| Hose Barb | 1/16 in. |
| Maximum Pressure | 2.7 bar |
| Product Type | Pump Tubing |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu